missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-thiocarboxamide, 97%

Manufacturer:  Alfa Aesar™ H54438MD

 View more versions of this product

Catalog No. AAH54438MD



Specifications

Specifications

Indole-3-thiocarboxamide
59108-90-6
MFCD01993552
PPWNZMJWESSLGE-UHFFFAOYSA-N
1H-indole-3-carbothioamide
12276962
97%
250mg
C9H8N2S
1h-indole-3-carbothioic acid amide
C1=CC=C2C(=C1)C(=CN2)C(=S)N
176.237
176.24

Chemical Identifiers

59108-90-6
176.237
PPWNZMJWESSLGE-UHFFFAOYSA-N
12276962
C1=CC=C2C(=C1)C(=CN2)C(=S)N
C9H8N2S
MFCD01993552
1h-indole-3-carbothioic acid amide
1H-indole-3-carbothioamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents