missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-3-lactic Acid, TCI America™

$131.39 - $711.95

Chemical Identifiers

CAS 1821-52-9
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00005642
InChI Key XGILAAMKEQUXLS-UHFFFAOYSA-N
Synonym 3-(3-Indolyl)lactic Acid, 2-Hydroxy-3-(3-indolyl)propionic Acid
PubChem CID 92904
ChEBI CHEBI:24813
IUPAC Name 2-hydroxy-3-(1H-indol-3-yl)propanoic acid
SMILES C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I0031100MG View Documents TCI America
I0031100MG
100mg Each for $131.39
 
I00311G View Documents TCI America
I00311G
1g Each for $711.95
 
Specifications

Chemical Identifiers

1821-52-9
205.213
XGILAAMKEQUXLS-UHFFFAOYSA-N
92904
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
C11H11NO3
MFCD00005642
3-(3-Indolyl)lactic Acid, 2-Hydroxy-3-(3-indolyl)propionic Acid
CHEBI:24813
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O

Specifications

White-Yellow
100mg
C11H11NO3
3-(3-Indolyl)lactic Acid, 2-Hydroxy-3-(3-indolyl)propionic Acid
C1=CC=C2C(=C1)C(=CN2)CC(C(=O)O)O
205.213
CHEBI:24813
Crystal-Powder at 20°C
Indole-3-lactic Acid
145°C
1821-52-9
MFCD00005642
XGILAAMKEQUXLS-UHFFFAOYSA-N
2-hydroxy-3-(1H-indol-3-yl)propanoic acid
92904
205.21
≥98.0% (HPLC,T)
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator