Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-glyoxylyl chloride, 98+%

Chemical Identifiers

Chemical Name or Material Indole-3-glyoxylyl chloride
CAS 22980-09-2
Molecular Formula C10H6ClNO2
Formula Weight 207.62
MDL Number MFCD00015460
UN Number UN2923
Beilstein 147693
Synonym alpha-Oxoindoleacetyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0090804

 
alfa aesar™
L0090804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0090809

 
alfa aesar™
L0090809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0090818

 
alfa aesar™
L0090818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.62
Chemical Name or Material Indole-3-glyoxylyl chloride
Assay ≥98%
Molecular Formula C10H6ClNO2
MDL Number MFCD00015460
CAS 22980-09-2
Synonym alpha-Oxoindoleacetyl chloride
Health Hazard 1 Danger
Melting Point ≈130°C
Odor Irritating
Health Hazard 2 GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2923
Beilstein 147693
Quantity 2g
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C