missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-carboxamidoxime, 97%

Manufacturer:  Alfa Aesar™ H5442903

 View more versions of this product

Catalog No. AAH5442903



Specifications

Specifications

Indole-3-carboxamidoxime
95649-37-9
MFCD00460181
3-indolecarboxylic acid amidoxime
C1=CC=C2C(=C1)C(=C(N)NO)C=N2
175.191
175.19
1g
C9H9N3O
4249031
DRCFXQBNDIRJCT-CLFYSBASSA-N
N-[(E)-amino(indol-3-ylidene)methyl]hydroxylamine
6097083
97%

Chemical Identifiers

95649-37-9
175.191
DRCFXQBNDIRJCT-CLFYSBASSA-N
6097083
C1=CC=C2C(=C1)C(=C(N)NO)C=N2
C9H9N3O
MFCD00460181
3-indolecarboxylic acid amidoxime
N-[(E)-amino(indol-3-ylidene)methyl]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents