missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-carboxaldoxime, 97%

Manufacturer:  Alfa Aesar™ H5475406

 View more versions of this product

Catalog No. AAH5475406



Specifications

Specifications

Indole-3-carboxaldoxime
2592-05-4
MFCD00022737
HFNHXOHLZXORGW-VOTSOKGWSA-N
N-[(Z)-indol-3-ylidenemethyl]hydroxylamine
5310834
97%
5g
C9H8N2O
1h-indole-3-carbaldehyde oxime
C1=CC=C2C(=C1)C(=CNO)C=N2
160.18
160.17

Chemical Identifiers

2592-05-4
160.18
HFNHXOHLZXORGW-VOTSOKGWSA-N
5310834
C1=CC=C2C(=C1)C(=CNO)C=N2
C9H8N2O
MFCD00022737
1h-indole-3-carbaldehyde oxime
N-[(Z)-indol-3-ylidenemethyl]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents