missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-carboxaldoxime, 97%

Manufacturer:  Alfa Aesar™ H5475403

 View more versions of this product

Catalog No. AAH5475403



Specifications

Specifications

Indole-3-carboxaldoxime
2592-05-4
MFCD00022737
HFNHXOHLZXORGW-VOTSOKGWSA-N
N-[(Z)-indol-3-ylidenemethyl]hydroxylamine
5310834
97%
1g
C9H8N2O
1h-indole-3-carbaldehyde oxime
C1=CC=C2C(=C1)C(=CNO)C=N2
160.18
160.17

Chemical Identifiers

2592-05-4
160.18
HFNHXOHLZXORGW-VOTSOKGWSA-N
5310834
C1=CC=C2C(=C1)C(=CNO)C=N2
C9H8N2O
MFCD00022737
1h-indole-3-carbaldehyde oxime
N-[(Z)-indol-3-ylidenemethyl]hydroxylamine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents