missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-3-carbinol, 97%, ACROS Organics™

Chemical Identifiers

CAS 700-06-1
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00005632
InChI Key IVYPNXXAYMYVSP-UHFFFAOYSA-N
Synonym 1h-indol-3-yl methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indole methanol, 3-indolemethanol, 3-indolylcarbinol, indinol, indole 3 carbinol, indole-3-carbinol, indole-3-methanol
PubChem CID 3712
ChEBI CHEBI:24814
IUPAC Name 1H-indol-3-ylmethanol
SMILES C1=CC=C2C(=C1)C(=CN2)CO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC122190050
SDS View Documents
Fisher Scientific Acros
122190050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC122190250
SDS View Documents
Thermo Fisher Scientific
122190250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

700-06-1
147.18
IVYPNXXAYMYVSP-UHFFFAOYSA-N
3712
1H-indol-3-ylmethanol
C9H9NO
MFCD00005632
1h-indol-3-yl methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indole methanol, 3-indolemethanol, 3-indolylcarbinol, indinol, indole 3 carbinol, indole-3-carbinol, indole-3-methanol
CHEBI:24814
C1=CC=C2C(=C1)C(=CN2)CO

Specifications

Authentic
Orange-Yellow to White
5g
96% min. (HPLC)
MFCD00005632
1h-indol-3-yl methanol, 1h-indole-3-methanol, 3-hydroxymethylindole, 3-indole methanol, 3-indolemethanol, 3-indolylcarbinol, indinol, indole 3 carbinol, indole-3-carbinol, indole-3-methanol
IVYPNXXAYMYVSP-UHFFFAOYSA-N
1H-indol-3-ylmethanol
3712
147.18
97%
Glass bottle
92.0°C to 94.0°C
700-06-1
C9H9NO
21, V,3, 45
Solubility in water: soluble. Other solubilities: soluble in ethanol,chloroform,benzene,toluene,,diethyl ether,petroleum ether
C1=CC=C2C(=C1)C(=CN2)CO
147.18
CHEBI:24814
Crystalline Powder or Flakes
Indole-3-carbinol, 99%
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning

EINECSNumber : 211-836-2

RTECSNumber : NL9483000

SDS
Product Certifications