Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-3-acetic acid hydrazide, 98%

$48.66 - $48.66

Chemical Identifiers

Chemical Name or Material Indole-3-acetic acid hydrazide
CAS 5448-47-5
Molecular Formula C10H11N3O
Formula Weight 189.22
MDL Number MFCD00005634
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1656203

 
alfa aesar™
A1656203
1g Each for $48.66
Description & Specifications

Specifications

Formula Weight 189.22
CAS 5448-47-5
Melting Point 142° to 145°C
Molecular Formula C10H11N3O
Quantity 1g
MDL Number MFCD00005634
RTECS Number NL3600000
Assay 98%
Chemical Name or Material Indole-3-acetic acid hydrazide