Learn More
Indole-3-acetic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1055636
Description
It is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is applied as plant growth hormone. is an inducer of plant cell elongation and division shown to cause uncontrolled growth.
Solubility
Soluble in ethanol (50 mg/ml), methanol, DMSO, and chloroform (sparingly). Insoluble in water.
Notes
Light sensitive. Store at -20°C. ncompatible with oxidizing agents.
Specifications
Indole-3-acetic acid | |
∼167°C (decomposition) | |
C10H9NO2 | |
500 g | |
Light sensitive | |
indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
2-(1H-indol-3-yl)acetic acid | |
802 | |
175.19 |
87-51-4 | |
171°C (339°F) | |
MFCD00005636 | |
143358 | |
14,4964 | |
Soluble in ethanol (50mg/ml),methanol,DMSO,and chloroform (sparingly). Insoluble in water. | |
OC(=O)CC1=CNC2=CC=CC=C12 | |
175.19 | |
CHEBI:16411 | |
≥98% |
Chemical Identifiers
87-51-4 | |
175.19 | |
SEOVTRFCIGRIMH-UHFFFAOYSA-N | |
802 | |
2-(1H-indol-3-yl)acetic acid |
C10H9NO2 | |
MFCD00005636 | |
indole-3-acetic acid, 3-indoleacetic acid, heteroauxin, indoleacetic acid, 1h-indole-3-acetic acid, 2-1h-indol-3-yl acetic acid, rhizopin, 1h-indol-3-ylacetic acid, indol-3-ylacetic acid, rhizopon a | |
CHEBI:16411 | |
OC(=O)CC1=CNC2=CC=CC=C12 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 201-748-2
RTECSNumber : NL3150000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only