missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indole-3-acetamide 98.0+%, TCI America™
$59.73 - $233.92
Chemical Identifiers
CAS | 879-37-8 |
---|---|
Molecular Formula | C10H10N2O |
Molecular Weight (g/mol) | 174.20 |
MDL Number | MFCD00005641 |
InChI Key | ZOAMBXDOGPRZLP-UHFFFAOYSA-N |
PubChem CID | 397 |
ChEBI | CHEBI:16031 |
IUPAC Name | 2-(1H-indol-3-yl)acetamide |
SMILES | NC(=O)CC1=CNC2=CC=CC=C12 |
Chemical Identifiers
879-37-8 | |
174.20 | |
ZOAMBXDOGPRZLP-UHFFFAOYSA-N | |
CHEBI:16031 | |
NC(=O)CC1=CNC2=CC=CC=C12 |
C10H10N2O | |
MFCD00005641 | |
397 | |
2-(1H-indol-3-yl)acetamide |
Specifications
879-37-8 | |
White-Yellow | |
MFCD00005641 | |
ZOAMBXDOGPRZLP-UHFFFAOYSA-N | |
2-(1H-indol-3-yl)acetamide | |
397 | |
174.20 | |
Crystalline Powder |
153°C | |
C10H10N2O | |
5 g | |
NC(=O)CC1=CNC2=CC=CC=C12 | |
174.20 | |
CHEBI:16031 | |
≥98.0% (HPLC,N) | |
Indole-3-acetamide |
Safety and Handling
TSCA : No