Learn More
Indole-2-carboxylic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 122200500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indole-2-carboxylic acid | |
1477-50-5 | |
Brown to White | |
98.5% min. (HPLC) | |
C9H7NO2 | |
50 g | |
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole | |
HCUARRIEZVDMPT-UHFFFAOYSA-N | |
1H-indole-2-carboxylic acid | |
72899 | |
99% |
99% | |
205.0°C to 209.0°C | |
Authentic | |
Glass bottle | |
MFCD00005611 | |
22, 61 | |
Solubility in water: insoluble. Other solubilities: soluble in ethanol,ether | |
OC(=O)C1=CC2=CC=CC=C2N1 | |
161.16 | |
161.16 | |
Crystalline Powder or Crystals |
Chemical Identifiers
1477-50-5 | |
161.16 | |
HCUARRIEZVDMPT-UHFFFAOYSA-N | |
72899 | |
OC(=O)C1=CC2=CC=CC=C2N1 |
C9H7NO2 | |
MFCD00005611 | |
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole | |
1H-indole-2-carboxylic acid |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 216-030-4
RUO – Research Use Only