Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-2-carboxylic acid, 98+%

$85.58 - $85.58

Chemical Identifiers

Chemical Name or Material Indole-2-carboxylic acid
CAS 1477-50-5
Molecular Formula C9H7NO2
Formula Weight 161.16
MDL Number MFCD00005611
Beilstein 124132
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1869009

  SDS
alfa aesar™
A1869009
10g Each for $85.58
Description & Specifications

Specifications

Formula Weight 161.16
RTECS Number NK7882812
Health Hazard 2 GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 10g
Health Hazard 1 Danger
MDL Number MFCD00005611
Molecular Formula C9H7NO2
Beilstein 124132
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
Melting Point ≈205°C (dec.)
CAS 1477-50-5
Chemical Name or Material Indole-2-carboxylic acid

Applications
Reactant for total synthesis of (±)-dibromophakellin and analogs, synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine, stereo selective preparation of renieramycin G analogs, preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization, Pd-catalyzed cyclization and N,N′-(pentane)diylbis[indolecarboxamide] and N,N′-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives.

Solubility
Soluble in clear yellow solution 5% in ethanol, dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.