missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1869018

 View more versions of this product

Catalog No. AAA1869018



Description

Description

Reactant for total synthesis of (±)-dibromophakellin and analogs, synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine, stereo selective preparation of renieramycin G analogs, preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization, Pd-catalyzed cyclization and N,N-(pentane)diylbis[indolecarboxamide] and N,N-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for total synthesis of (±)-dibromophakellin and analogs, synthesis of the pyrrolizidine alkaloid (±)-trachelanthamidine, stereo selective preparation of renieramycin G analogs, preparation of spirooxoindolepyrrolidines via reduction of indole-2-carboxylic acid followed by oxidation, condensation, reduction, amidation and Kharasch radical cyclization, Pd-catalyzed cyclization and N,N′-(pentane)diylbis[indolecarboxamide] and N,N′-[phenylenebis(methylene)]bis[indolecarboxamide] derivatives.

Solubility
Soluble in clear yellow solution 5% in ethanol, dimethyl sulfoxide and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Indole-2-carboxylic acid
∼205°C (decomposition)
MFCD00005611
124132
Soluble in clear yellow solution 5% in ethanol,dimethyl sulfoxide and methanol.
OC(=O)C1=CC2=CC=CC=C2N1
161.16
161.16
1477-50-5
C9H7NO2
50 g
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
HCUARRIEZVDMPT-UHFFFAOYSA-N
1H-indole-2-carboxylic acid
72899
≥98%

Chemical Identifiers

1477-50-5
161.16
HCUARRIEZVDMPT-UHFFFAOYSA-N
72899
OC(=O)C1=CC2=CC=CC=C2N1
C9H7NO2
MFCD00005611
indole-2-carboxylic acid, 2-carboxyindole, 2-indolecarboxylic acid, indol-2-carboxylic acid, 1h-indolecarboxylic acid, indole-2-carboxylate, 2-indolylformic acid, 2-indole carboxylic acid, 1h-indol-2-carbons, carboxyindole
1H-indole-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312-H315-H319-H335

EINECSNumber : 216-030-4

RTECSNumber : NK7882812

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only