missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-2-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H54638MD

 View more versions of this product

Catalog No. AAH54638MD



Description

Description

Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

Solubility
Soluble in Methanol.

Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
Specifications

Specifications

Indole-2-carboxaldehyde
138°C to 142°C
19005-93-7
MFCD03001425
Soluble in Methanol.
C1=CC=C2C(=C1)C=C(N2)C=O
145.161
145.16
Air Sensitive
250mg
C9H7NO
1h-indole-2-carboxaldehyde
SBNOTUDDIXOFSN-UHFFFAOYSA-N
1H-indole-2-carbaldehyde
96389
97%

Chemical Identifiers

19005-93-7
145.161
SBNOTUDDIXOFSN-UHFFFAOYSA-N
96389
C1=CC=C2C(=C1)C=C(N2)C=O
C9H7NO
MFCD03001425
1h-indole-2-carboxaldehyde
1H-indole-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents