missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indole-2-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H5463806

 View more versions of this product

Catalog No. AAH5463806



Description

Description

Applications
Indole-2-carboxaldehyde is a useful starter in indole and natural product chemistry.

Solubility
Soluble in Methanol.

Notes
Air sensitive. Strong oxidizers and strong reducing agents are incompatible. Store at room temperature.
Specifications

Specifications

Indole-2-carboxaldehyde
5g
C9H7NO
1h-indole-2-carboxaldehyde
C1=CC=C2C(=C1)C=C(N2)C=O
145.161
145.16
138°C to 142°C
19005-93-7
MFCD03001425
SBNOTUDDIXOFSN-UHFFFAOYSA-N
1H-indole-2-carbaldehyde
96389
97%

Chemical Identifiers

19005-93-7
145.161
SBNOTUDDIXOFSN-UHFFFAOYSA-N
96389
C1=CC=C2C(=C1)C=C(N2)C=O
C9H7NO
MFCD03001425
1h-indole-2-carboxaldehyde
1H-indole-2-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

Documents