Please login to your online account to display your discounted pricing

Individual Aroclors, SPEX CertiPrep™

$64.86 - $64.86

Chemical Identifiers

Chemical Name or Material Aroclor 1262 (High Level)
CAS 37324-23-5
CAS Min % 0.1
CAS Max % 0.1
CAS 110-54-3
CAS Min % 99.9
CAS Max % 99.9
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PCB-1262H

 
spex certiprep llc
PCB1262H
2mL amber prescored glass ampule 1mL Each for $64.86
Description & Specifications

Specifications

Quantity 1mL
Methods For US EPA Methods 505, 508, 625, 8082, 8270C
Solvent or Matrix Hexane
Chemical Name or Material Aroclor 1262 (High Level)
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
DOT Information Nonhazardous
Packaging 2mL amber prescored glass ampule
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Recommended Storage Ambient
CAS Max % 99.9
CAS Min % 0.1
CAS 37324-23-5
CAS Max % 0.1
CAS 110-54-3
CAS Min % 99.9

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Ambient