missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Form: approx. 5.74g/m
Supplier: Thermo Scientific Chemicals 000470G5
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver-White | |
7440-74-6 | |
99.999% (Metals basis) | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
[In] | |
114.82 | |
CHEBI:30430 | |
Wire |
Indium | |
Silvery-white wire | |
In | |
5 m | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
Odorless |
Chemical Identifiers
7440-74-6 | |
114.82 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
5359967 | |
indium |
In | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
CHEBI:30430 | |
[In] |
Safety and Handling
EINECSNumber : 231-180-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only