missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium wire, 2.0mm (0.08 in.) dia., Puratronic™, 99.998+% (metals basis)

Manufacturer:  Alfa Aesar™ 11461G5

 View more versions of this product

Catalog No. AA11461G5



Specifications

Specifications

Indium wire
5m
7440-74-6
In
indio
[In]
Odorless
5359967
Wire
Puratronic™
≈22.95g/m
2.0mm (0.08 in.) dia.
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
≥99.998%

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents