missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium Wire, 2.0mm (0.08 in.) dia., 99.99% (Metals basis), Alfa Aesar™

Form: Approx.22.95g/m

Manufacturer:  Alfa Aesar 43265G1

 View more versions of this product

Catalog No. AA43265G1



Specifications

Specifications

Indium
7440-74-6
7.31g/cm3 at 20°C
In
indio
[In]
Odorless
5359967
Wire, 2mm (0.08 in.) dia.
1m
Silvery-white wire
99.99% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Silver-White

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Documents