Please login to your online account to display your discounted pricing

Indium wire, 1.0mm (0.04in) dia, Puratronic™, 99.998+% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Indium wire
CAS 7440-74-6
Name Note 1.0mm (0.04 in.) dia, Puratronic™
MDL Number MFCD00134048
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA11462G5

 
alfa aesar
11462G5
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5m
Chemical Name or Material Indium wire
TSCA TSCA
Density ≈5.74g/m
Name Note 1.0mm (0.04 in.) dia, Puratronic™
Assay ≥99.998%
Assay Percent Notes (metals basis)
MDL Number MFCD00134048
CAS 7440-74-6
Odor Odorless