missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium wire, 1.0mm (0.04 in.) dia., annealed, Puratronic™, 99.998+% (metals basis)

Manufacturer:  Alfa Aesar™ 46661G5

 View more versions of this product

Catalog No. AA46661G5



Specifications

Specifications

Indium wire
5m
7440-74-6
≥99.998%
In
APFVFJFRJDLVQX-UHFFFAOYSA-N
[In]
114.818
CHEBI:30430
≈5.74g/m
1.0mm (0.04 in.) dia., Annealed
Puratronic™
(metals basis)
MFCD00134048
Odorless
indium
5359967
Wire

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
CHEBI:30430
[In]
In
MFCD00134048
5359967
indium
Safety and Handling

Safety and Handling

RTECSNumber : NL1050000

TSCA : TSCA

SDS Documents