missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium Wire, 0.5mm (0.02 in.) dia., Puratronic™, >99.998% (Metals basis), Alfa Aesar™

Form: Approx.1.43g/m

Manufacturer:  Alfa Aesar 11463G9

 View more versions of this product

Catalog No. AA11463G9



Specifications

Specifications

Indium
7440-74-6
>99.998% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Silver-White
25m
Silvery-white wire
In
indio
[In]
Odorless
5359967
Wire, 0.5mm (0.02 in.) dia.

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Documents