missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium Standard (Trace ICP/ICP-MS Grade), SPEX CertiPrep™

$126.80 - $126.80

Chemical Identifiers

Chemical Name or Material Indium Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
CLIN21AM Spex Certiprep Llc
SPXCLIN21AM
Triple-leached LDPE bottle 30mL Each for $126.80
Specifications

Description

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Description & Specifications

Chemical Identifiers

Indium Standard

Specifications

30mL
Triple-leached LDPE bottle
2% HNO3
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Indium Standard

Safety and Handling

Causes skin irritation, serious eye irritation

DOTInformation : Nonhazardous

Recommended Storage : Ambient