missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium Standard, 1000μg/mL in 2-5% HNO3 (Assurance Grade), SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$200.89 - $365.49
Chemical Identifiers
CAS | 7440-74-6 |
---|---|
Molecular Formula | In |
Molecular Weight (g/mol) | 114.818 |
InChI Key | APFVFJFRJDLVQX-UHFFFAOYSA-N |
PubChem CID | 5359967 |
ChEBI | CHEBI:30430 |
IUPAC Name | indium |
SMILES | [In] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
In | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-74-6 | |
114.818 | |
5359967 | |
indium |
In | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
CHEBI:30430 | |
[In] |
Specifications
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation | |
0.1,2.0,97.9 | |
In | |
[In] | |
114.818 | |
CHEBI:30430 | |
Triple-leached LDPE bottle | |
2% HNO3 | |
Indium Standard |
Matrix Concentration 1000μg/mL 2-5% HNO3 |
|
7440-74-6,7697-37-2,7732-18-5 | |
0.1,2.0,97.9 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
Plasma | |
Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
125mL |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only