Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium spheres, 0.5mm (0.02 in.) dia., 99.99% (metals basis)

Chemical Identifiers

Chemical Name or Material Indium
CAS 7440-74-6
Name Note 0.5mm (0.02 in.) dia.
MDL Number MFCD00134048
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4724506

 
alfa aesar™
4724506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4724518

 
alfa aesar™
4724518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4724530

 
alfa aesar™
4724530
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Indium
TSCA TSCA
Name Note 0.5mm (0.02 in.) dia.
Physical Form Spheres
Assay 99.99%
Assay Percent Notes (metals basis)
MDL Number MFCD00134048
CAS 7440-74-6
Odor Odorless
Quantity 5g