missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium shot, tear drop, 4mm (0.2 in.), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1102609

 View more versions of this product

Catalog No. AA1102609



Specifications

Specifications

Indium shot
4mm (0.2 in.)
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
99.9%
10g
7440-74-6
In
indio
[In]
Odorless
5359967
Teardrop Shot

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents