missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium shot, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 1102730

 View more versions of this product

Catalog No. AA1102730



Specifications

Specifications

Indium
7440-74-6
7.31g/cm3 at 20°C
In
indio
[In]
Odorless
5359967
Shots, Tear Drop, 4mm (0.2 in.)
250g
Silvery-white shot
99.99% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Silver-White

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

EINECSNumber : 231-180-0

Documents