missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium shot, 5mm (0.2 in.) & down, Puratronic™, 99.9999+% (metals basis)

Manufacturer:  Alfa Aesar™ 1061418

 View more versions of this product

Catalog No. AA1061418



Specifications

Specifications

Indium shot
7440-74-6
In
indio
[In]
Odorless
5359967
Shot
≥99.9999%
50g
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
5mm (0.2 in.) & down
Puratronic™

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents