missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium rod, 2.54mm (0.10 in.) dia., 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 43249BV

 View more versions of this product

Catalog No. AA43249BV



Specifications

Specifications

Indium rod
30cm
7440-74-6
In
indio
[In]
Odorless
5359967
Rod
≈0.37g/cm
2.54mm (0.10 in.) dia.
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
99.99%

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents