Learn More
Indium powder, -100 mesh, 99.99% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04498622
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
156.6°C | |
2072°C | |
(metals basis) | |
MFCD00134048 | |
100 g | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
114.82 | |
−100 Mesh | |
Powder |
Indium powder | |
7440-74-6 | |
7.31 g/mL | |
In | |
UN3089 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
[In] | |
114.82 | |
CHEBI:30430 | |
99.99% | |
Odorless |
Chemical Identifiers
7440-74-6 | |
114.82 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
5359967 | |
indium |
In | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
CHEBI:30430 | |
[In] |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H228-H302+H312+H332
DOTInformation : Hazard Class: 4.1; Packaging Group: II
EINECSNumber : 231-180-0
RTECSNumber : NL1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only