Learn More
Indium powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04512214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver-White | |
156.6°C | |
2072°C | |
7.31 | |
In | |
25 g | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
indium | |
5359967 | |
114.82 | |
Odorless |
Indium | |
7440-74-6 | |
Silver white powder | |
99.9% (Metals basis) | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
[In] | |
114.82 | |
CHEBI:30430 | |
-100 Mesh | |
Powder |
Chemical Identifiers
7440-74-6 | |
114.82 | |
APFVFJFRJDLVQX-UHFFFAOYSA-N | |
5359967 | |
indium |
In | |
MFCD00134048 | |
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic | |
CHEBI:30430 | |
[In] |
Safety and Handling
Keep away from sources of ignition - No Smoking.
P210-P240-P241-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c
H228-H302+H312+H332
DOTInformation : DOT Class: 4.1; Packaging Group: II
EINECSNumber : 231-180-0
RTECSNumber : NL1050000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only