missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium Powder, -100 mesh, 99.9% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4512206

 View more versions of this product

Catalog No. AA4512206



Specifications

Specifications

Indium
7440-74-6
7.31g/cm3 at 20°C
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
Odorless
indium
5359967
114.82
Silver-White
5g
Silver white powder
99.9% (Metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
[In]
114.818
CHEBI:30430
Powder

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

Keep away from sources of ignition - No Smoking.

H228: Flammable solid.

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-180-0

SDS Documents