Learn More
Indium oxide, tin doped, polymeric precursor, Oxide ≈20 wt%, Thermo Scientific Chemicals
Preparation of transparent electrical conducting electrodes
Supplier: Thermo Scientific Chemicals 04222909
Description
It is used in the preparation of transparent electrical conducting electrodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium oxide | |
1312-43-2 | |
10 g | |
MFCD00011060 | |
13°C (55°F) | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
diindium(3+) trioxidandiide | |
Oxide ≈20 wt% |
Tin doped, polymeric precursor | |
Viscous Liquid | |
In2O3 | |
UN1993 | |
Soluble in hexane and toluene; Insoluble in alcohols | |
[O--].[O--].[O--].[In+3].[In+3] | |
277.63 |
Chemical Identifiers
1312-43-2 | |
277.63 | |
PJXISJQVUVHSOJ-UHFFFAOYSA-N | |
[O--].[O--].[O--].[In+3].[In+3] |
In2O3 | |
MFCD00011060 | |
diindium(3+) trioxidandiide |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H225-H319-H336-H372
DOTInformation : Hazard Class: 3; Packaging Group: II
TSCA : Yes
RUO – Research Use Only