Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9999+% (metals basis)

Chemical Identifiers

Chemical Name or Material Indium ingot
CAS 7440-74-6
Purity Grade Puratronic™
MDL Number MFCD00134048
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1061818

 
alfa aesar™
1061818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Indium ingot
TSCA TSCA
Purity Grade Puratronic™
Physical Form Polycrystalline Ingot
Assay ≥99.9999%
Assay Percent Notes (metals basis)
MDL Number MFCD00134048
CAS 7440-74-6
Odor Odorless
Quantity 50g