missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9999% (metals basis)

$765.03 - $3,844.42

Chemical Identifiers

CAS 7440-74-6
Molecular Formula In
Molecular Weight (g/mol) 114.818
MDL Number MFCD00134048
InChI Key APFVFJFRJDLVQX-UHFFFAOYSA-N
Synonym indio, indium, elemental, ingot, polycrystalline, puratronic, powder, powder,-100 mesh, shot, shot, 5mm 0.2in & down, puratronic, shot, tear drop, 4mm 0.2in, unii-045a6v3vfx, wire, 0.5mm 0.02in dia, puratronic
PubChem CID 5359967
ChEBI CHEBI:30430
IUPAC Name indium
SMILES [In]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10617MX
View Documents
Alfa Aesar™
10617MX
5 x 12.5g
Each for $874.31
 
AA1061718
View Documents
Alfa Aesar™
1061718
50g
Each for $765.03
 
AA1061790
View Documents
Alfa Aesar™
1061790
5 x 50g
Each for $3,844.42
 
Description

Description

Specifications

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio, indium, elemental, ingot, polycrystalline, puratronic, powder, powder,-100 mesh, shot, shot, 5mm 0.2in & down, puratronic, shot, tear drop, 4mm 0.2in, unii-045a6v3vfx, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]

Specifications

5 x 12.5g
In
indio, indium, elemental, ingot, polycrystalline, puratronic, powder, powder,-100 mesh, shot, shot, 5mm 0.2in & down, puratronic, shot, tear drop, 4mm 0.2in, unii-045a6v3vfx, wire, 0.5mm 0.02in dia, puratronic
[In]
Odorless
5359967
114.82
99.9999%
Indium ingot
7440-74-6
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Polycrystalline Ingot
Puratronic™
(metals basis)
Safety and Handling

Safety and Handling

EINECSNumber : 231-180-0

RTECSNumber : NL1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS