missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 10617TH

 View more versions of this product

Catalog No. AA10617TH



Specifications

Specifications

Indium ingot
7440-74-6
In
indio
[In]
Odorless
5359967
114.82
99.9999%
12.5g
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Polycrystalline Ingot
Puratronic™

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

RTECSNumber : NL1050000

TSCA : TSCA

Documents