missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 1061790

 View more versions of this product

Catalog No. AA1061790



Specifications

Specifications

Indium ingot
7440-74-6
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
[In]
Odorless
5359967
114.82
99.9999%
5 x 50g
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Polycrystalline Ingot
Puratronic™

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

RTECSNumber : NL1050000

TSCA : TSCA

Documents