missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, polycrystalline, Puratronic™, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 1061690

 View more versions of this product

Catalog No. AA1061690



Specifications

Specifications

Indium ingot
7440-74-6
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
indium
114.818
CHEBI:30430
Polycrystalline Ingot
Puratronic™
2072°C
5 x 50g
7.310g/mL
In
indio
[In]
Odorless
5359967
114.818
99.9995%
156.6°C

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indio
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents