missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium ingot, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1472018

 View more versions of this product

Catalog No. AA1472018



Specifications

Specifications

Indium ingot
7440-74-6
In
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
Odorless
indium
5359967
Soft Ingot
White
50g
(metals basis)
MFCD00134048
APFVFJFRJDLVQX-UHFFFAOYSA-N
[In]
114.818
CHEBI:30430
99.999%

Chemical Identifiers

7440-74-6
114.818
APFVFJFRJDLVQX-UHFFFAOYSA-N
5359967
indium
In
MFCD00134048
indium, elemental, unii-045a6v3vfx, powder, shot, powder,-100 mesh, indio, shot, tear drop, 4mm 0.2in, ingot, polycrystalline, puratronic, shot, 5mm 0.2in & down, puratronic, wire, 0.5mm 0.02in dia, puratronic
CHEBI:30430
[In]
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents