Learn More
Indium(III) trifluoromethanesulfonate, 99% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 4013118
Description
Indium(III) trifluoromethanesulfonate is used as a Lewis acid catalyst in organic synthesis and as a co-catalyst in organometallic catalysis. It is also used as a reactant in the preparation of stable indium bacteriochlorins and decahydroquinoline-type toxins through intramolecular hetero Diels-Alder reactions. Further, it is used in the synthesis of benzoxazoles through cyclocondensations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium(III) trifluoromethanesulfonate | |
128008-30-0 | |
50 g | |
MFCD00144478 | |
indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t | |
UCYRAEIHXSVXPV-UHFFFAOYSA-K | |
indium(3+);trifluoromethanesulfonate | |
2734680 | |
≤99% |
(decomposition) | |
Powder | |
C3F9InO9S3 | |
UN3261 | |
Slightly soluble in hydrocarbons and diethyl ether. Insoluble in water | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3] | |
562.008 | |
562.02 | |
Hygroscopic |
Chemical Identifiers
128008-30-0 | |
562.008 | |
UCYRAEIHXSVXPV-UHFFFAOYSA-K | |
2734680 | |
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[In+3] |
C3F9InO9S3 | |
MFCD00144478 | |
indium iii trifluoromethanesulfonate, indium triflate, indium trifluoromethanesulphonate, indium 3+ tritriflate, indium 3+ ; trifluoromethanesulfonate, indium iii triflate, indium iii trifluoromethanesulphonate, acmc-209bda, in tfa 3, ksc491q8t | |
indium(3+);trifluoromethanesulfonate |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only