Please login to your online account to display your discounted pricing

Indium(III) trifluoromethanesulfonate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC368540050

 View more versions of this product

Catalog No. AC368540050

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder and Granules
Formula Weight 562.03
Quantity 5g
Color Off-white
Infrared Spectrum Authentic
CAS 128008-30-0
Chemical Name or Material Indium(III) trifluoromethanesulfonate
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C3F9InO9S3
MDL Number MFCD00144478
Synonym Indium(III) triflate
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Elemental Analysis 97.5% min. (ex Fluor)

Specifications

Chemical Name or Material Indium(III) trifluoromethanesulfonate
Assay Percent Range 98%
Molecular Formula C3F9InO9S3
Formula Weight 562.03
MDL Number MFCD00144478
Synonym Indium(III) triflate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

SDS & Safety Documents