missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) sulfide, 99.995% (metals basis)

Manufacturer:  Alfa Aesar™ 4483604

 View more versions of this product

Catalog No. AA4483604



Specifications

Specifications

Indium (III) sulfide
(metals basis)
MFCD00011061
Powder
Like rotten eggs
Red to Orange
2g
12030-24-9
In2S3
325.83
99.995%
Moisture Sensitive
1050°C

Chemical Identifiers

12030-24-9
In2S3
MFCD00011061
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents