Learn More
Indium(III) sulfide, 99.98% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04483504
Description
Indium(III) sulfide acts as mid band gap semiconductor. It is an important material for optoelectronic and photovoltaic applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium (III) sulfide | |
12030-24-9 | |
2 g | |
In2S3 | |
Insoluble in water. | |
[S--].[S--].[S--].[In+3].[In+3] | |
325.82 | |
99.98% | |
Moisture sensitive |
1050°C | |
Powder | |
(metals basis) | |
MFCD00011061 | |
SIXIBASSFIFHDK-UHFFFAOYSA-N | |
diindium(3+) trisulfanediide | |
325.83 | |
Like rotten eggs |
Chemical Identifiers
12030-24-9 | |
325.82 | |
SIXIBASSFIFHDK-UHFFFAOYSA-N | |
[S--].[S--].[S--].[In+3].[In+3] |
In2S3 | |
MFCD00011061 | |
diindium(3+) trisulfanediide |
Safety and Handling
GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H332-H315-H319-H335
EINECSNumber : 234-742-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only