missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium(III) sulfate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01071118
Description
Indium(III) sulfate is used Chemical synthesis, hardening agent in gold electroplating baths. It is also used to prepare other indium containing substances such as copper indium selenide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Indium(III) sulfate hydrate | |
13464-82-9 | |
50 g | |
In2O12S3 | |
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german | |
XGCKLPDYTQRDTR-UHFFFAOYSA-H | |
indium(3+);trisulfate | |
26044 | |
99.999% |
250°C (decomposition) | |
Lump/Powder | |
(metals basis) | |
MFCD00011059 | |
Soluble in water and ethanol. Insoluble in basic salts. | |
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O | |
517.80 | |
517.83 (Anhydrous) | |
Puratronic™ |
Chemical Identifiers
13464-82-9 | |
517.80 | |
XGCKLPDYTQRDTR-UHFFFAOYSA-H | |
26044 | |
[In+3].[In+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O |
In2O12S3 | |
MFCD00011059 | |
indium sulfate, indisulfat, indium sulphate, indium iii sulfate, diindium tris sulphate, indium trisulfate, diindium trisulfate, indium sequisulfate, sulfuric acid, indium salt, indisulfat german | |
indium(3+);trisulfate |
Safety and Handling
EINECSNumber : 236-689-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only