missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium(III) selenide, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8828014

 View more versions of this product

Catalog No. AA8828014



Specifications

Specifications

Indium(III) selenide
99.99% (Metal basis)
MFCD00016147
466.52
Odorless
25g
12056-07-4
In2Se3
Silver gray pieces
3 to 6mm Pieces
Silver to Gray

Chemical Identifiers

12056-07-4
In2Se3
MFCD00016147
Safety and Handling

Safety and Handling

H301-H331-H373-H400-H410: Toxic if swallowed. Toxic if inhaled. May cause damage to organs through prolonged or repeated exposure. Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

EINECSNumber : 235-016-9

SDS Documents