missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) phosphide, polycrystalline lump, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ A1550914

 View more versions of this product

Catalog No. AAA1550914



Description

Description

Applications
InP is used in high-power and high-frequency electronics because of its superior electron velocity. It also has a direct bandgap, making it useful for optoelectronics devices like laser diodes. It is also used as a substrate for epitaxial indium gallium arsenide based opto-electronic devices.

Solubility
Insoluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
Specifications

Specifications

Indium(III) phosphide
(metals basis)
MFCD00016153
indium phosphide, indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, indium iii phosphide, dsstox_cid_11444, dsstox_rid_78875
P#[In]
145.792
145.79
99.99%
25g
22398-80-7
InP
14,4953
GPXJNWSHGFTCBW-UHFFFAOYSA-N
indiganylidynephosphane
31170
Polycrystalline Lump
1070°C

Chemical Identifiers

22398-80-7
145.792
GPXJNWSHGFTCBW-UHFFFAOYSA-N
31170
P#[In]
InP
MFCD00016153
indium phosphide, indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, indium iii phosphide, dsstox_cid_11444, dsstox_rid_78875
indiganylidynephosphane
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NL1800000

TSCA : TSCA

Documents