missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Indium(III) phosphide, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 1397406

 View more versions of this product

Catalog No. AA1397406



Specifications

Specifications

Indium(III) phosphide
(metals basis)
MFCD00016153
indium phosphide, indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, indium iii phosphide, dsstox_cid_11444, dsstox_rid_78875
P#[In]
145.792
145.79
99.9999%
1070°C
22398-80-7
InP
14,4953
GPXJNWSHGFTCBW-UHFFFAOYSA-N
indiganylidynephosphane
31170
Pieces
4.81g/mL
5g

Chemical Identifiers

22398-80-7
145.792
GPXJNWSHGFTCBW-UHFFFAOYSA-N
31170
P#[In]
InP
MFCD00016153
indium phosphide, indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, indium iii phosphide, dsstox_cid_11444, dsstox_rid_78875
indiganylidynephosphane
Safety and Handling

Safety and Handling

GHS H Statement
H351
Suspected of causing cancer.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : NL1800000

TSCA : TSCA

Documents