Learn More
Indium(III) phosphide, 99.999% (metals basis), Thermo Scientific Chemicals
$340.89 - $1624.27
Chemical Identifiers
CAS | 22398-80-7 |
---|---|
Molecular Formula | InP |
Molecular Weight (g/mol) | 145.792 |
MDL Number | MFCD00016153 |
InChI Key | GPXJNWSHGFTCBW-UHFFFAOYSA-N |
Synonym | indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 |
PubChem CID | 31170 |
IUPAC Name | indiganylidynephosphane |
SMILES | P#[In] |
Description
The water soluble InP/ZnS core/shell QDs are a safer alternative to CdSe/ZnS QDs for biological applications, by comparing their toxicity in vitro (cell culture) and in vivo.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
22398-80-7 | |
145.792 | |
GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
31170 | |
P#[In] |
InP | |
MFCD00016153 | |
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
indiganylidynephosphane |
Specifications
1070°C | |
1 g | |
MFCD00016153 | |
indium phosphide, indium iii phosphide, indium monophosphide, unii-sd36lg60g1, ccris 8799, phosphinidyneindium, dsstox_cid_11444, dsstox_rid_78875 | |
GPXJNWSHGFTCBW-UHFFFAOYSA-N | |
indiganylidynephosphane | |
31170 | |
99.999% | |
Indium(III) phosphide |
22398-80-7 | |
InP | |
14,4953 | |
Insoluble in water. | |
P#[In] | |
145.792 | |
145.79 | |
4.81 g/mL | |
(metals basis) |
Safety and Handling
GHS H Statement
H351
Suspected of causing cancer.
P201-P202-P260-P264b-P270-P281-P308+P313-P501c
H350-H361f-H372
EINECSNumber : 244-959-5
RTECSNumber : NL1800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only