missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Indium(III) oxide, 99.9997%, (trace metal basis), ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC193170050

 View more versions of this product

Catalog No. AC193170050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

for ultra trace analysis
Specifications

Specifications

Indium(III) oxide
81.0 to 84.3% (In) (Complexometry)
In2O3
Solubility in water: insoluble
277.63
99.9997%
Glass bottle
3ppm max.
5g
1312-43-2
In2O3
15,4992
277.63
Powder
Trace Metal Basis
Solubility in water: insoluble.
Yellow

Chemical Identifiers

1312-43-2
In2O3
277.63
Safety and Handling

Safety and Handling

EINECSNumber : 215-193-9

SDS Documents