missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Indium(III) oxide, 99.9997%, (trace metal basis), Thermo Scientific™
Chemical Identifiers
CAS | 1312-43-2 |
---|---|
Molecular Formula | In2O3 |
Molecular Weight (g/mol) | 277.63 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC193170050
|
Thermo Scientific Chemicals
193170050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC193170250
|
Thermo Scientific Chemicals
193170250 |
25g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for ultra trace analysisChemical Identifiers
1312-43-2 | |
In2O3 | |
277.63 |
Specifications
1312-43-2 | |
81.0 to 84.3% (In) (Complexometry) | |
In2O3 | |
Solubility in water: insoluble | |
277.63 | |
Trace Metal Basis | |
Indium(III) oxide |
5g | |
In2O3 | |
15,4992 | |
277.63 | |
99.9997% | |
Glass bottle |
Safety and Handling
EINECSNumber : 215-193-9
RUO â Research Use Only